2,2-Dimethyl-N-(6-methylpyridin-2-yl)propanamide
Catalog # APOSOR310943-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.258 g/mol
Storage Temperature:Ambient
CAS Number:86847-79-2
Formula:C₁₁H₁₆N₂O
MDL Number:MFCD00996301