3,5-Dichloro-4-formylphenyl acetate
Catalog # APOSOR310629-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.048 g/mol
Melting Pt:119…121 °C
Storage Temperature:Ambient
CAS Number:1582770-02-2
Formula:C₉H₆Cl₂O₃
MDL Number:MFCD27980615