
3,5-Dichloro-4-(hydroxymethyl)phenol
Catalog # APOSOR310627-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.027 g/mol
Melting Pt:194…196 °C
:GHS07
Storage Temperature:Ambient
CAS Number:720679-54-9
Formula:C₇H₆Cl₂O₂
MDL Number:MFCD06797936