Order Entry
Export
ContactUsLinkComponent
5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine
5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine
Catalog # APOSOR310544-1G
Supplier:  Apollo Scientific
CAS Number:  
5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine
Catalog # APOSOR310544-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.693 g/mol
Storage Temperature:Ambient
CAS Number:848462-68-0
Formula:C₁₀H₇ClN₂OS
MDL Number:MFCD26393360