N-Phenylquinolin-4-amine
Catalog # APOSOR310414-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.269 g/mol
Storage Temperature:Ambient
CAS Number:30696-07-2
Formula:C₁₅H₁₂N₂
MDL Number:MFCD00958207