


1,1-Diethylpropargylamine
Catalog # APOSOR310413-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:111.185 g/mol
Density:0.824 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2733
ADR:3+8,II
:GHS02
:GHS05
:GHS07
CAS Number:3234-64-8
Formula:C₇H₁₃N
MDL Number:MFCD00008057