Order Entry
Export
ContactUsLinkComponent
Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate
Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate
Catalog # APOSOR310380-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate
Catalog # APOSOR310380-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.293 g/mol
Storage Temperature:Ambient
CAS Number:120596-96-5
Formula:C₈H₁₀O₃S₂
MDL Number:MFCD18800140