Order Entry
Export
ContactUsLinkComponent
(2S)-2,3-Dimethylbutane-1,3-diol
(2S)-2,3-Dimethylbutane-1,3-diol
Catalog # APOSOR310330-500MG
Supplier:  Apollo Scientific
CAS Number:  
(2S)-2,3-Dimethylbutane-1,3-diol
Catalog # APOSOR310330-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :118.174 g/mol
Storage Temperature:Ambient
CAS Number:73295-12-2
Formula:C₆H₁₄O₂
MDL Number:MFCD10574870