Order Entry
Export
ContactUsLinkComponent
(3,4-Diaminophenyl)methanol
Irritant
(3,4-Diaminophenyl)methanol
Catalog # APOSOR310306-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(3,4-Diaminophenyl)methanol
Catalog # APOSOR310306-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :138.167 g/mol
Melting Pt:97…99 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:63189-98-0
Formula:C₇H₁₀N₂O
MDL Number:MFCD00173951