4-(Chloromethyl)-1,2-dinitrobenzene
Catalog # APOSOR310304-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.579 g/mol
Storage Temperature:Ambient
CAS Number:89692-62-6
Formula:C₇H₅ClN₂O₄
MDL Number:MFCD13689020