Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl hydrazinecarbodithioate
Methyl hydrazinecarbodithioate
Methyl hydrazinecarbodithioate
Irritant
Methyl hydrazinecarbodithioate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl hydrazinecarbodithioate
Supplier:
Apollo Scientific
About this item
CAS Number:
5397-03-5
:
Formula:
C₂H₆N₂S₂
MDL Number:
MFCD00463356
:
122.212 g/mol
:
GHS07
Storage Temperature:
Ambient