4,6-Dibromoquinoline
Catalog # APOSOR309460-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.951 g/mol
Storage Temperature:Ambient
CAS Number:927801-13-6
Formula:C₉H₅Br₂N
MDL Number:MFCD08436999