Order Entry
Export
ContactUsLinkComponent
(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Catalog # APOSOR309409-1G
Supplier:  Apollo Scientific
CAS Number:  
(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Catalog # APOSOR309409-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.262 g/mol
Storage Temperature:Ambient
CAS Number:185034-18-8
Formula:C₁₀H₉N₃O₂S
MDL Number:MFCD05721268