Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
APOSOR309387-1G
:
Apollo Scientific
:
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
APOSOR309387-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
252.107 g/mol
:
GHS07
:
Ambient
:
1367706-85-1
:
C₁₁H₁₀BrNO
:
MFCD21647766