3-Bromo-2,8-dimethyl-4-hydroxyquinoline
Catalog # APOSOR309372-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.107 g/mol
Storage Temperature:Ambient
CAS Number:909208-88-4
Formula:C₁₁H₁₀BrNO
MDL Number:MFCD18157848