

6-Bromo-3,4-dichloroquinoline
Catalog # APOSOR309336-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:128…129 °C
:276.945 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:927801-17-0
Formula:C₉H₄BrCl₂N
MDL Number:MFCD08437001