Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,6,8-trimethylquinoline
4-Bromo-2,6,8-trimethylquinoline
Catalog # APOSOR309294-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,6,8-trimethylquinoline
Catalog # APOSOR309294-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.134 g/mol
Storage Temperature:Ambient
CAS Number:1070879-60-5
Formula:C₁₂H₁₂BrN
MDL Number:MFCD11505145