4-Bromo-5,8-dichloroquinoline
Catalog # APOSOR309287-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.945 g/mol
Storage Temperature:Ambient
CAS Number:1070879-38-7
Formula:C₉H₄BrCl₂N
MDL Number:MFCD11505120