4-Bromo-5,7-dimethylquinoline
Catalog # APOSOR309286-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:236.108 g/mol
Storage Temperature:Ambient
CAS Number:1070879-33-2
Formula:C₁₁H₁₀BrN
MDL Number:MFCD11505114