4-Bromo-7-methylquinoline
Catalog # APOSOR309285-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.081 g/mol
Storage Temperature:Ambient
CAS Number:1070879-24-1
Formula:C₁₀H₈BrN
MDL Number:MFCD08063191