7-Bromo-2-hydroxy-3-phenylquinoline
Catalog # APOSOR309276-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:300.15 g/mol
Storage Temperature:Ambient
CAS Number:861872-50-6
Formula:C₁₅H₁₀BrNO
MDL Number:MFCD09997966