7-Bromo-2-chloro-3-phenylquinoline
Catalog # APOSOR309274-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:318.596 g/mol
Storage Temperature:Ambient
CAS Number:85274-44-8
Formula:C₁₅H₉BrClN
MDL Number:MFCD09997964