Order Entry
Export
ContactUsLinkComponent
2-Chloro-6-methoxy-3-phenylquinoline
2-Chloro-6-methoxy-3-phenylquinoline
Catalog # APOSOR309261-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-methoxy-3-phenylquinoline
Catalog # APOSOR309261-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.726 g/mol
Storage Temperature:Ambient
CAS Number:85274-57-3
Formula:C₁₆H₁₂ClNO
MDL Number:MFCD09998016