Order Entry
Export
ContactUsLinkComponent
2,6-Dichloroquinoline-3-methanol
2,6-Dichloroquinoline-3-methanol
Catalog # APOSOR309256-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloroquinoline-3-methanol
Catalog # APOSOR309256-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.075 g/mol
Storage Temperature:Ambient
CAS Number:1017429-35-4
Formula:C₁₀H₇Cl₂NO
MDL Number:MFCD09997988