5,8-Dimethylquinoline-3-carboxylic acid
Catalog # APOSOR307990-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.221 g/mol
Storage Temperature:Ambient
CAS Number:763893-29-4
Formula:C₁₂H₁₁NO₂
MDL Number:MFCD09787777