5,7-Dimethylquinoline-3-carboxylic acid
Catalog # APOSOR307988-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.221 g/mol
Storage Temperature:Ambient
CAS Number:948293-95-6
Formula:C₁₂H₁₁NO₂
MDL Number:MFCD09787770