Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dichloroquinoline-3-carboxylic acid
5,7-Dichloroquinoline-3-carboxylic acid
5,7-Dichloroquinoline-3-carboxylic acid
5,7-Dichloroquinoline-3-carboxylic acid
APOSOR307986-1G
:
Apollo Scientific
:
5,7-Dichloroquinoline-3-carboxylic acid
APOSOR307986-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
242.058 g/mol
:
Ambient
:
948293-83-2
:
C₁₀H₅Cl₂NO₂
:
MFCD09787766