2,6-Dichloroquinoline-3-carbonitrile
Catalog # APOSOR307917-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.058 g/mol
Storage Temperature:Ambient
CAS Number:948291-61-0
Formula:C₁₀H₄Cl₂N₂
MDL Number:MFCD09787669