3-(2-Chloroethyl)-2,7-dichloroquinoline
Catalog # APOSOR307897-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260.547 g/mol
Storage Temperature:Ambient
CAS Number:948294-54-0
Formula:C₁₁H₈Cl₃N
MDL Number:MFCD09787623