Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,8-Dimethylquinoline hydrochloride
2,8-Dimethylquinoline hydrochloride
2,8-Dimethylquinoline hydrochloride
2,8-Dimethylquinoline hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
2,8-Dimethylquinoline hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
53826-01-0
:
Formula:
C₁₁H₁₁N·ClH
MDL Number:
MFCD16987423
:
193.673 g/mol
Storage Temperature:
Ambient