4-[(4-Amino-phenylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSOR307784-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.415 g/mol
Storage Temperature:Ambient
CAS Number:1159976-33-6
Formula:C₁₇H₂₇N₃O₂
MDL Number:MFCD12026438