1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone hydrochloride
Catalog # APOSOR307762-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:240.729 g/mol
Storage Temperature:Ambient
CAS Number:1226815-20-8
Formula:C₁₂H₁₆N₂O·ClH
MDL Number:MFCD15530198