1-(3,3-Dimethyl-6-nitroindolin-1-yl)ethanone
Catalog # APOSOR307761-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.251 g/mol
Storage Temperature:Ambient
CAS Number:453562-68-0
Formula:C₁₂H₁₄N₂O₃
MDL Number:MFCD07636645