Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
APOSOR307627-1G
:
Apollo Scientific
:
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
APOSOR307627-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
220.096 g/mol
:
159…160 °C
:
Ambient
:
3259
:
8,III
:
GHS05
:
787615-23-0
:
C₉H₁₁Cl₂NO
:
MFCD06227486