Order Entry
Export
Orders LinkContactUsLinkComponent
4,7-Dibromoquinoline

4,7-Dibromoquinoline

Supplier:   Apollo Scientific
4,7-Dibromoquinoline
Supplier:  Apollo Scientific
Certificates
4,7-Dibromoquinoline

4 Options Available Below

Product Details & Documents
Synonyms:  4,7-Dibromo-1-azanaphthalene
  • CAS Number:700871-88-1
  • Formula:C₉H₅Br₂N
  • MDL Number:MFCD08063188
  • Molecular Weight:286.951 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 95,60 €
    95,60 €
    1 * 100 mg
       95,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 118,00 €
    118,00 €
    1 * 250 mg
       118,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 238,00 €
    238,00 €
    1 * 1 g
       238,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 830,00 €
    830,00 €
    1 * 5 g
       830,00 €

    Specifications:
    New Product
    New Product