4,8-Dibromoquinoline
Catalog # APOSOR307501-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.951 g/mol
Storage Temperature:Ambient
CAS Number:1070879-31-0
Formula:C₉H₅Br₂N
MDL Number:MFCD08063207