Order Entry
Export
ContactUsLinkComponent
1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
Irritant
1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
Catalog # APOSOR307297-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(1-Ethoxyethyl)-3,4-diiodo-1H-pyrazole
Catalog # APOSOR307297-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :391.976 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1639858-70-0
Formula:C₇H₁₀I₂N₂O
MDL Number:MFCD30721064