Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Diiodo-1H-pyrazole
3,4-Diiodo-1H-pyrazole 95%
3,4-Diiodo-1H-pyrazole 95%
Irritant
3,4-Diiodo-1H-pyrazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,4-Diiodo-1H-pyrazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
6645-70-1
:
Formula:
C₃H₂I₂N₂
MDL Number:
MFCD26793484
:
319.87 g/mol
:
GHS07
Storage Temperature:
Ambient