
2-(3,4-Dimethoxypyridin-2-yl)-3,4-dimethoxypyridine
Catalog # APOSOR307231-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:276.288 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:101664-55-5
Formula:C₁₄H₁₆N₂O₄
MDL Number:MFCD25976502