
(R,R)-Cyclohex-4-ene-1,2-dicarboxylic acid
Catalog # APOSOR307025-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:170.163 g/mol
Melting Pt:167…168 °C
:GHS07
Storage Temperature:Ambient
CAS Number:15573-40-7
Formula:C₈H₁₀O₄
MDL Number:MFCD09757670