Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethylmorpholine
3,5-Dimethylmorpholine
3,5-Dimethylmorpholine
Corrosive
Irritant
3,5-Dimethylmorpholine
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
3,5-Dimethylmorpholine
Supplier:
Apollo Scientific
About this item
CAS Number:
123-57-9
:
Formula:
C₆H₁₃NO
MDL Number:
MFCD00065032
:
115.174 g/mol
UN:
1993
ADR:
3,III
:
GHS05
:
GHS07
Storage Temperature:
Ambient