Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
Irritant
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1H-Pyrrolo[2,3-b]pyridine-2,3-dione
Supplier:
Apollo Scientific
About this item
CAS Number:
5654-95-5
:
Formula:
C₇H₄N₂O₂
MDL Number:
MFCD09999197
:
148.119 g/mol
:
GHS07
Storage Temperature:
Ambient