3-Iodo-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR305572-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.021 g/mol
Storage Temperature:Ambient
CAS Number:633328-40-2
Formula:C₆H₄IN₃
MDL Number:MFCD10696864