
4-Chloro-2-methyl[1,3]oxazolo[5,4-c]pyridine
Catalog # APOSOR305553-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:168.58 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1354831-15-4
Formula:C₇H₅ClN₂O
MDL Number:MFCD24848274