5,7-Dichloro-1H-pyrrolo[2,3-c]pyridine
Catalog # APOSOR305545-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:187.026 g/mol
Storage Temperature:Ambient
CAS Number:1001412-41-4
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD11845567