Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Irritant
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
79173-38-9
:
Formula:
C₇H₈N₄
MDL Number:
MFCD00456552
:
148.165 g/mol
:
GHS07
Storage Temperature:
Ambient