Order Entry
Export
ContactUsLinkComponent
6-Bromo-1H-pyrazolo[4,3-c]pyridine
Irritant
6-Bromo-1H-pyrazolo[4,3-c]pyridine
Catalog # APOSOR305335-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-1H-pyrazolo[4,3-c]pyridine
Catalog # APOSOR305335-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :198.02 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1206973-12-7
Formula:C₆H₄BrN₃
MDL Number:MFCD17010106