Order Entry
Export
ContactUsLinkComponent
4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one 95%
4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one 95%
Catalog # APOSOR305276-250MG
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one 95%
Catalog # APOSOR305276-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :202.037 g/mol
Storage Temperature:Ambient
CAS Number:74572-30-8
Formula:C₈H₅Cl₂NO
MDL Number:MFCD13177812