7-Bromo-5-chloro-1,2-benzoxazole
Catalog # APOSOR305029-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:232.462 g/mol
Storage Temperature:Ambient
CAS Number:1260903-12-5
Formula:C₇H₃BrClNO
MDL Number:MFCD11847343