
4,4',4''-[(Benzene-1,3,5-triyl)triethyne-2,1-diyl]tripyridine
Catalog # APOSOR305007-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:381.428 g/mol
Melting Pt:237 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:168289-78-9
Formula:C₂₇H₁₅N₃
MDL Number:MFCD25541760